siRNA Details
| Gene Symbol: TPT1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055081.1 | - | 1 | 133 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000434849.2 | - | 1 | 133 | Chr. 13 - 45374934 - 45378616 [+] on Build GRCh38 | |

TPT1-AS1
Genomic Map