siRNA Details
| Gene Symbol: LINC00689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125809.1 | - | 1 | 1816 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00689| Gene Symbol: LINC00689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK125809.1 | - | 1 | 1816 | ||