siRNA Details
Gene Symbol: FAM209B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC014951.1 | - | 1 | 416 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM209B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC014951.1 | - | 1 | 416 |