siRNA Details
| Gene Symbol: LOC101927410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110777.1 | 3 | 1633 | |||
| GenBank mRNA | AK056572.1 | - | 3 | 1633 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000562202.1 | - | 3 | 1633 | Chr. 18 - 10661933 - 10666887 [+] on Build GRCh38 | |

LOC101927410
Genomic Map