siRNA Details
| Gene Symbol: SIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005267100.2 | XP_005267157.1 | 12 | 8197 | ||
| XM_017011197.1 | XP_016866686.1 | 12 | 7929 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SIM1| Gene Symbol: SIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005267100.2 | XP_005267157.1 | 12 | 8197 | ||
| XM_017011197.1 | XP_016866686.1 | 12 | 7929 | |||