siRNA Details
| Gene Symbol: CDC42-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF086337.1 | - | 1 | 334 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | W75995.1 | - | 335 | Chr. - - [-] on Build | ||

CDC42-IT1
Genomic Map