siRNA Details
| Gene Symbol: CTD-2201E9.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109935.1 | 1, 2 | 83 | |||
| GenBank mRNA | AK057486.1 | - | 1, 2 | 83 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB025218.1 | - | 1, 2 | 83 | Chr. 5 - 9511333 - 9516960 [+] on Build GRCh38 | |

CTD-2201E9.1
Genomic Map