siRNA Details
| Gene Symbol: CTD-2201E9.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109935.1 | 3 | 1377 | |||
| GenBank mRNA | AK057486.1 | - | 3 | 1377 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTD-2201E9.1| Gene Symbol: CTD-2201E9.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109935.1 | 3 | 1377 | |||
| GenBank mRNA | AK057486.1 | - | 3 | 1377 | ||