siRNA Details
| Gene Symbol: SMARCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003601.3 | NP_003592.3 | 24 | 6751 | ||
| GenBank mRNA | AK123439.1 | - | 1 | 822 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMARCA5| Gene Symbol: SMARCA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003601.3 | NP_003592.3 | 24 | 6751 | ||
| GenBank mRNA | AK123439.1 | - | 1 | 822 | ||