siRNA Details
| Gene Symbol: LOC102546229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105044.1 | 3 | 379 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK057158.1 | - | 2 | 521 | Chr. 8 - 109298598 - 109316562 [+] on Build GRCh38 | |

LOC102546229
Genomic Map