siRNA Details
| Gene Symbol: LOC105378499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092127.1 | - | 1 | 306 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105378499| Gene Symbol: LOC105378499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092127.1 | - | 1 | 306 | ||