siRNA Details
| Gene Symbol: LOC105370792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135681.1 | 2, 3 | 404 | |||
| GenBank mRNA | AK057625.1 | - | 1, 2 | 2536 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105370792| Gene Symbol: LOC105370792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135681.1 | 2, 3 | 404 | |||
| GenBank mRNA | AK057625.1 | - | 1, 2 | 2536 | ||