siRNA Details
Gene Symbol: LINC01004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039981.2 | 1 | 8083 | |||
GenBank mRNA | AK092446.1 | - | 1 | 822 | ||
AK123595.1 | - | 1 | 1839 | |||
BC041969.1 | - | 1 | 844 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01004 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039981.2 | 1 | 8083 | |||
GenBank mRNA | AK092446.1 | - | 1 | 822 | ||
AK123595.1 | - | 1 | 1839 | |||
BC041969.1 | - | 1 | 844 |