siRNA Details
| Gene Symbol: FUBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011519172.2 | XP_011517474.1 | 17 | 1910 | ||
| GenBank mRNA | BC128537.1 | - | 1 | 535 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FUBP3| Gene Symbol: FUBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011519172.2 | XP_011517474.1 | 17 | 1910 | ||
| GenBank mRNA | BC128537.1 | - | 1 | 535 | ||