siRNA Details
| Gene Symbol: PRSS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003619.3 | NP_003610.2 | 13 | 4174 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRSS12| Gene Symbol: PRSS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003619.3 | NP_003610.2 | 13 | 4174 | ||