siRNA Details
| Gene Symbol: D21S2088E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040254.1 | 5 | 580 | |||
| GenBank mRNA | AY063451.1 | - | 5 | 580 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D21S2088E| Gene Symbol: D21S2088E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040254.1 | 5 | 580 | |||
| GenBank mRNA | AY063451.1 | - | 5 | 580 | ||