siRNA Details
Gene Symbol: LINC00307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038855.1 | 3 | 332 | |||
GenBank mRNA | AY063455.1 | - | 3 | 332 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000451410.1 | - | 3 | 332 | Chr. 21 - 30209151 - 30211783 [-] on Build GRCh38 |
