siRNA Details
| Gene Symbol: LOC101928700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110829.1 | 2, 3 | 319 | |||
| GenBank mRNA | BC026117.1 | - | 2, 3 | 324 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG720584.1 | - | 2, 3 | 323 | Chr. 7 - 147082866 - 147097605 [-] on Build GRCh38 | |

LOC101928700
Genomic Map