siRNA Details
Gene Symbol: LOC414300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121211.1 | 1 | 238 | |||
GenBank mRNA | BC032033.1 | - | 1 | 231 | ||
HY054313.1 | - | 1 | 238 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC414300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121211.1 | 1 | 238 | |||
GenBank mRNA | BC032033.1 | - | 1 | 231 | ||
HY054313.1 | - | 1 | 238 |