siRNA Details
| Gene Symbol: LOC414300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121211.1 | 1 | 503 | |||
| GenBank mRNA | BC032033.1 | - | 1 | 496 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC414300| Gene Symbol: LOC414300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121211.1 | 1 | 503 | |||
| GenBank mRNA | BC032033.1 | - | 1 | 496 | ||