siRNA Details
Gene Symbol: LOC101928865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120564.1 | 4 | 1477 | |||
GenBank mRNA | BC039514.1 | - | 4 | 1477 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120564.1 | 4 | 1477 | |||
GenBank mRNA | BC039514.1 | - | 4 | 1477 |