siRNA Details
| Gene Symbol: LOC101927164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110668.1 | 3 | 1064 | |||
| GenBank mRNA | BC039513.1 | - | 3 | 1064 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927164| Gene Symbol: LOC101927164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110668.1 | 3 | 1064 | |||
| GenBank mRNA | BC039513.1 | - | 3 | 1064 | ||