siRNA Details
| Gene Symbol: LOC101927164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110668.1 | 1 | 119 | |||
| GenBank mRNA | BC039513.1 | - | 1 | 119 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM465960.1 | - | 1 | 117 | Chr. 1 - 224219615 - 224227574 [+] on Build GRCh38 | |

LOC101927164
Genomic Map