siRNA Details
| Gene Symbol: WDR86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006715966.2 | XP_006716029.1 | 5 | 1418 | ||
| GenBank mRNA | AK096225.1 | - | 1 | 381 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDR86| Gene Symbol: WDR86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006715966.2 | XP_006716029.1 | 5 | 1418 | ||
| GenBank mRNA | AK096225.1 | - | 1 | 381 | ||