siRNA Details
| Gene Symbol: RASGRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF370414.1 | - | 1 | 1389 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CX760929.1 | - | 1 | 518 | Chr. 15 - 78979042 - 78979771 [+] on Build GRCh38 | |

RASGRF1
Genomic Map