siRNA Details
| Gene Symbol: FAM135B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC038357.1 | - | 2 | 586 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG723289.1 | - | 2 | 591 | Chr. 8 - 138441278 - 138477461 [-] on Build GRCh38 | |

FAM135B
Genomic Map