siRNA Details
| Gene Symbol: LOC284933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038917.1 | 5 | 1120 | |||
| GenBank mRNA | BC036359.1 | - | 5 | 1121 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC284933| Gene Symbol: LOC284933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038917.1 | 5 | 1120 | |||
| GenBank mRNA | BC036359.1 | - | 5 | 1121 | ||