siRNA Details
Gene Symbol: LOC100422212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104294.1 | 1 | 2085 | |||
GenBank mRNA | BC036607.1 | - | 1 | 2080 | ||
BC047397.1 | - | 1 | 2086 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100422212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104294.1 | 1 | 2085 | |||
GenBank mRNA | BC036607.1 | - | 1 | 2080 | ||
BC047397.1 | - | 1 | 2086 |