siRNA Details
| Gene Symbol: LOC100422212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC036607.1 | - | 1 | 2055 | ||
| BC047397.1 | - | 1 | 2061 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100422212| Gene Symbol: LOC100422212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC036607.1 | - | 1 | 2055 | ||
| BC047397.1 | - | 1 | 2061 | |||