siRNA Details
| Gene Symbol: ENTPD1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC028619.1 | - | 4 | 1551 | ||
| Gene Symbol: CC2D2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK022203.1 | - | 1 | 1127 | ||

ENTPD1-AS1
Genomic Map