siRNA Details
| Gene Symbol: ASB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012303.1 | XP_016867792.1 | 5 | 1703 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK129691.1 | - | 1 | 2144 | Chr. 7 - 95542145 - 95544542 [+] on Build GRCh38 | |

ASB4
Genomic Map