siRNA Details
Gene Symbol: LOC101928661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125860.1 | 3 | 1846 | |||
GenBank mRNA | BC034912.1 | - | 3 | 1845 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125860.1 | 3 | 1846 | |||
GenBank mRNA | BC034912.1 | - | 3 | 1845 |