siRNA Details
| Gene Symbol: LOC105378910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC044921.2 | - | 1 | 1239 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105378910| Gene Symbol: LOC105378910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC044921.2 | - | 1 | 1239 | ||