siRNA Details
| Gene Symbol: GNRH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017027823.1 | XP_016883312.1 | 1 | 856 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GNRH2| Gene Symbol: GNRH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017027823.1 | XP_016883312.1 | 1 | 856 | ||