siRNA Details
| Gene Symbol: LOC105378146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136248.1 | 1 | 529 | |||
| NR_136249.1 | 2 | 144 | ||||
| GenBank mRNA | BC044924.2 | - | 1 | 491 | ||
| BG771988.1 | - | 1 | 492 | |||
| BX088781.1 | - | 2 | 95 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105378146| Gene Symbol: LOC105378146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136248.1 | 1 | 529 | |||
| NR_136249.1 | 2 | 144 | ||||
| GenBank mRNA | BC044924.2 | - | 1 | 491 | ||
| BG771988.1 | - | 1 | 492 | |||
| BX088781.1 | - | 2 | 95 | |||