siRNA Details
Gene Symbol: LOC105378146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136248.1 | 1 | 502 | |||
GenBank mRNA | BC044924.2 | - | 1 | 464 | ||
BG771988.1 | - | 1 | 465 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105378146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136248.1 | 1 | 502 | |||
GenBank mRNA | BC044924.2 | - | 1 | 464 | ||
BG771988.1 | - | 1 | 465 |