siRNA Details
| Gene Symbol: JAZF1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034097.1 | 4 | 1073 | |||
| GenBank mRNA | AK293540.1 | - | 4 | 1073 | ||
| BC043567.1 | - | 2 | 1023 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

JAZF1-AS1| Gene Symbol: JAZF1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034097.1 | 4 | 1073 | |||
| GenBank mRNA | AK293540.1 | - | 4 | 1073 | ||
| BC043567.1 | - | 2 | 1023 | |||