siRNA Details
| Gene Symbol: ZFP92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005274652.3 | XP_005274709.1 | 6 | 5772 | ||
| XM_011531115.2 | XP_011529417.1 | 5 | 5723 | |||
| XM_017029281.1 | XP_016884770.1 | 7 | 6121 | |||
| GenBank mRNA | AK057819.1 | - | 1 | 2088 | ||

ZFP92
Genomic Map