siRNA Details
| Gene Symbol: LOC283484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090861.1 | - | 1 | 2228 | ||
| AK127899.1 | - | 1 | 1378 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283484| Gene Symbol: LOC283484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK090861.1 | - | 1 | 2228 | ||
| AK127899.1 | - | 1 | 1378 | |||