siRNA Details
| Gene Symbol: MYHAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125367.1 | 11 | 2157 | |||
| GenBank mRNA | AK097500.1 | - | 11 | 2145 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYHAS| Gene Symbol: MYHAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125367.1 | 11 | 2157 | |||
| GenBank mRNA | AK097500.1 | - | 11 | 2145 | ||