siRNA Details
| Gene Symbol: ZNF101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300949.1 | NP_001287878.1 | 4 | 4457 | ||
| NM_033204.3 | NP_149981.2 | 4 | 4585 | |||
| GenBank mRNA | AK123904.1 | - | 1 | 2174 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX105661.1 | - | 1 | 431 | Chr. 19 - 19682973 - 19683509 [+] on Build GRCh38 | |

ZNF101
Genomic Map