siRNA Details
| Gene Symbol: ZNF101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300949.1 | NP_001287878.1 | 4 | 4237 | ||
| NM_033204.3 | NP_149981.2 | 4 | 4365 | |||
| GenBank mRNA | AK123904.1 | - | 1 | 1954 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF101| Gene Symbol: ZNF101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300949.1 | NP_001287878.1 | 4 | 4237 | ||
| NM_033204.3 | NP_149981.2 | 4 | 4365 | |||
| GenBank mRNA | AK123904.1 | - | 1 | 1954 | ||