siRNA Details
| Gene Symbol: GCSAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005247272.2 | XP_005247329.1 | 1 | 448 | ||
| XM_011512629.1 | XP_011510931.1 | 1 | 448 | |||
| XM_011512630.1 | XP_011510932.1 | 1 | 448 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK128276.1 | - | 1 | 448 | Chr. 3 - 112130159 - 112133274 [-] on Build GRCh38 | |

GCSAM
Genomic Map