siRNA Details
| Gene Symbol: SGK494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174103.1 | NP_001167574.1 | 12 | 2284 | ||
| GenBank mRNA | AK056265.1 | - | 1 | 93 | ||
| BC001565.1 | - | 1 | 1483 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SGK494| Gene Symbol: SGK494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174103.1 | NP_001167574.1 | 12 | 2284 | ||
| GenBank mRNA | AK056265.1 | - | 1 | 93 | ||
| BC001565.1 | - | 1 | 1483 | |||