siRNA Details
| Gene Symbol: SGK494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC001565.1 | - | 1 | 218 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF377868.1 | - | 1 | 401 | Chr. 17 - 28610673 - 28611121 [-] on Build GRCh38 | |

SGK494
Genomic Map