siRNA Details
| Gene Symbol: PSMD13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK092845.1 | - | 1 | 1595 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | H50723.1 | - | 1 | 52 | Chr. 11 - 252330 - 252619 [+] on Build GRCh38 | |

PSMD13
Genomic Map