siRNA Details
| Gene Symbol: FLJ11710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021772.1 | - | 1 | 2050 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ11710| Gene Symbol: FLJ11710 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021772.1 | - | 1 | 2050 | ||