siRNA Details
| Gene Symbol: LOC102724659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126000.1 | 3 | 1019 | |||
| GenBank mRNA | BC034298.1 | - | 3 | 1019 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724659| Gene Symbol: LOC102724659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126000.1 | 3 | 1019 | |||
| GenBank mRNA | BC034298.1 | - | 3 | 1019 | ||