siRNA Details
Gene Symbol: LINC01247 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110251.1 | 1 | 1939 | |||
GenBank mRNA | BC044918.1 | - | 1 | 1948 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000448901.1 | - | 1 | 1939 | Chr. 2 - 6366010 - 6375422 [-] on Build GRCh38 |
