siRNA Details
| Gene Symbol: RAB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB073648.1 | - | 1 | 539 | ||
| AK055409.1 | - | 1 | 570 | |||
| AK094685.1 | - | 1 | 1416 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB3C| Gene Symbol: RAB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB073648.1 | - | 1 | 539 | ||
| AK055409.1 | - | 1 | 570 | |||
| AK094685.1 | - | 1 | 1416 | |||